Order Entry
Czech Republic
ContactUsLinkComponent
cis-1-Iodo-2-(heptafluoropropyl)cyclohexane
cis-1-Iodo-2-(heptafluoropropyl)cyclohexane
Catalog # APOSPC8758-100MG
Supplier:  Apollo Scientific
CAS Number:  
cis-1-Iodo-2-(heptafluoropropyl)cyclohexane
Catalog # APOSPC8758-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :378.07 g/mol
Boiling Pt:50 °C/1.3mmHg
Density:1.804 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:7589-43-7
Formula:C₉H₁₀F₇I
MDL Number:MFCD01862054