
1-Bromo-4-fluoro-2-iodobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:202865-72-3
:
Formula:C₆H₃BrFI
MDL Number:MFCD00042578
:300.9 g/mol
Boiling Pt:235 °C
Density:2.3 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together