2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:937602-49-8
:
Formula:C₁₀H₁₀F₃NOS
MDL Number:MFCD09027148
:249.26 g/mol
Melting Pt:78…80 °C
Storage Temperature:Ambient
Frequently Bought Together