
4-Bromo-2,2-difluoro-1,3-benzodioxole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:144584-66-7
:
Formula:C₇H₃BrF₂O₂
MDL Number:MFCD01631385
:237 g/mol
Boiling Pt:78…79 °C/12mmHg
Density:1.721 g/cm³ (25 °C)
Flash Pt:90.6 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together