Order Entry
Czech Republic
ContactUsLinkComponent
{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol
{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol
Catalog # APOSPC8593-500MG
Supplier:  Apollo Scientific
CAS Number:  
{2-[3-Fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazol-5-yl}methanol
Catalog # APOSPC8593-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :291.27 g/mol
Melting Pt:159…161 °C
Storage Temperature:Ambient
CAS Number:317319-27-0
Formula:C₁₂H₉F₄NOS
MDL Number:MFCD07369416