Order Entry
Czech Republic
ContactUsLinkComponent
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Catalog # APOSPC8590-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[3-Fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Catalog # APOSPC8590-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :291.23 g/mol
Melting Pt:178…180 °C
Storage Temperature:Ambient
CAS Number:937602-43-2
Formula:C₁₁H₅F₄NO₂S
MDL Number:MFCD09027138