
2-[(4-Fluorobenzyl)sulphanyl]ethylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1170404-43-9
:
Formula:C₉H₁₃ClFNS
MDL Number:MFCD09027133
:221.73 g/mol
Melting Pt:167…169 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together