Ethyl-2-[3-fluoro-4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:937602-41-0
:
Formula:C₁₃H₉F₄NO₂S
MDL Number:MFCD09027134
Melting Pt:111…113 °C
:319.28 g/mol
Storage Temperature:Ambient
Frequently Bought Together