1-[2-(4-Fluoroanilino)-1,3-thiazol-5-yl]-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952183-71-0
:
Formula:C₁₁H₉N₂OFS
MDL Number:MFCD09817444
Melting Pt:217…219 °C
Storage Temperature:Ambient
Frequently Bought Together