1-[2-(4-Fluoroanilino)-4-methyl-1,3-thiazol-5-yl]-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952183-69-6
:
Formula:C₁₂H₁₁N₂OFS
MDL Number:MFCD01804834
Melting Pt:165…167 °C
Storage Temperature:Ambient
Frequently Bought Together