2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:910037-17-1
:
Formula:C₁₂H₈F₃N₃
MDL Number:MFCD09817514
:251.21 g/mol
Melting Pt:101…103 °C
Storage Temperature:Ambient
Frequently Bought Together