Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Octafluoroadipamide
Octafluoroadipamide 97%
Octafluoroadipamide 97%
Octafluoroadipamide 97%
APOSPC8549-1G
:
Apollo Scientific
:
Octafluoroadipamide 97%
APOSPC8549-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
288.1 g/mol
:
236…238 °C
:
Ambient
:
1759
:
8,III
:
GHS05
:
GHS07
:
355-66-8
:
C₆H₄F₈N₂O₂
:
MFCD00446345