3,4-Difluorophenylzinc bromide 0,5 M in THF
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:737797-28-3
:
Formula:C₆H₃BrF₂Zn
MDL Number:MFCD02260154
:258.371 g/mol
:GHS02
Storage Temperature:Ambient
Frequently Bought Together