
3-Bromo-5-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:453565-90-7
:
Formula:C₈H₄BrF₃O₃
MDL Number:MFCD04116041
:285.02 g/mol
Melting Pt:87…89 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together