4-[(4-Fluorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
Catalog # APOSPC8488-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.28 g/mol
Melting Pt:151…152 °C
Storage Temperature:Ambient
CAS Number:952182-59-1
Formula:C₁₃H₈FNO₃S
MDL Number:MFCD09027121