Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride 95+%
Corrosive
Irritant
2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride 95+%
Catalog # APOSPC8484-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2,6-Difluoro-N-hydroxybenzenecarboximidoylchloride 95+%
Catalog # APOSPC8484-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :191.57 g/mol
Melting Pt:104…106 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:118591-69-8
Formula:C₇H₄ClF₂NO
MDL Number:MFCD09027155