
2,6-Difluorobenzenecarbaldehyde oxime
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19064-16-5
:
Formula:C₇H₅F₂NO
MDL Number:MFCD01764489
:157.12 g/mol
Melting Pt:114…116 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together