Ethyl-4-(2-chloro-6-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:299920-90-4
:
Formula:C₁₅H₁₅ClFNO₃
MDL Number:MFCD00664286
Boiling Pt:200…202 °C
Melting Pt:200…202 °C
:311.74 g/mol
Storage Temperature:Ambient
Frequently Bought Together