
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-nitrophenyl)acetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:213994-29-7
:
Formula:C₁₄H₇ClF₃N₃O₂
MDL Number:MFCD00172215
Melting Pt:110…112 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together