
2-Bromo-3'-fluoro-5'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:202664-38-8
:
Formula:C₉H₅BrF₄O
MDL Number:MFCD03094507
Melting Pt:49…52 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together