2-Chloro-5-fluoro-4-methylpyridine 97+%
Catalog # APOSPC8383-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:145.5600 g/mol
Storage Temperature:Ambient
CAS Number:881891-83-4
Formula:C₆H₅ClFN
MDL Number:MFCD09864695