
5-Bromo-2-fluoro-4-methylpyridine
Catalog # APOSPC8382-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:190.01 g/mol
Density:1.62 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:864830-16-0
Formula:C₆H₅BrFN
MDL Number:MFCD09839213