O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate 98%
Catalog # APOSPC8351-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:359.21 g/mol
Storage Temperature:Ambient
CAS Number:265651-18-1
Formula:C₉H₁₆F₆N₃O₃P
MDL Number:MFCD01863753