
2,6-Difluoro-4-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:123843-66-3
:
Formula:C₈H₅F₂NO
MDL Number:MFCD03094498
:169.13 g/mol
Melting Pt:53…57 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together