
2-Fluoro-5-methoxybenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367-83-9
:
Formula:C₈H₇FO₃
MDL Number:MFCD00272566
Melting Pt:146…150 °C
:170.14 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together