
2-Chloro-3-ethoxy-6-fluorophenylboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957120-93-3
:
Formula:C₈H₉BClFO₃
MDL Number:MFCD09800880
:218.4200 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together