3-Bromo-N,N-diethyl-5-(trifluoromethyl)benzenesulphonamide
Catalog # APOSPC8080-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:44…45 °C
Storage Temperature:Refrigerator
CAS Number:951885-25-9
Formula:C₁₁H₁₃BrF₃NO₂S
MDL Number:MFCD09800988