3-{2-[4-(Trifluoromethyl)piperidino]ethyl}benzenecarbaldehyde
Catalog # APOSPC7996-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.31 g/mol
Storage Temperature:Ambient
CAS Number:1000339-89-8
Formula:C₁₅H₁₈F₃NO
MDL Number:MFCD09864684