Tris(4-fluorophenyl)phosphine 97%
:18437-78-0
:
:C₁₈H₁₂F₃P
:MFCD00013553
:316.26 g/mol
:80…81 °C
:Ambient
Frequently Bought Together
Water, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylacetamide 99%, Reagent Grade
: Honeywell Chemicals
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
: Thermo Fisher Scientific
1,2-DIMETHOXYETHANE 1 * 1 L
: Merck
Cyclohexane for pesticide residue analysis
: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals