Order Entry
Czech Republic
ContactUsLinkComponent
Tris(3-fluorophenyl)phosphine 97%
Irritant
Tris(3-fluorophenyl)phosphine 97%
Catalog # APOSPC7886B-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Tris(3-fluorophenyl)phosphine 97%
Catalog # APOSPC7886B-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:62…63 °C
 :316.26 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:23039-94-3
Formula:C₁₈H₁₂F₃P
MDL Number:MFCD00015722