Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-5-fluoro-4-methoxyaniline 98%
3-Bromo-5-fluoro-4-methoxyaniline 98%
Catalog # APOSPC7874-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-5-fluoro-4-methoxyaniline 98%
Catalog # APOSPC7874-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.04 g/mol
Storage Temperature:Ambient
CAS Number:875664-44-1
Formula:C₇H₇BrFNO
MDL Number:MFCD08062384