
1-[2-(Trifluoromethyl)phenyl]propanol 98+%
Catalog # APOSPC7868-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:204.19 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:878572-13-5
Formula:C₁₀H₁₁F₃O
MDL Number:MFCD08062381