
Perfluoro(4-methyl-3,6-dioxaoct-7-ene)sulphonyl fluoride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16090-14-5
:
Formula:C₇F₁₄O₄S
MDL Number:MFCD00798138
:446.12 g/mol
Boiling Pt:135 °C
Density:1.7 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together