
1,3,5-Trifluoro-2-nitrobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:315-14-0
:
Formula:C₆H₂F₃NO₂
MDL Number:MFCD00014687
:177.08 g/mol
Boiling Pt:182…185 °C
Melting Pt:44318 °C
Density:1.514 g/cm³ (25 °C)
Flash Pt:78 °C
EINECS:206-248-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together