

2-Trifluoromethyl-2-propanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:507-52-8
:
Formula:C₄H₇F₃O
MDL Number:MFCD00014391
:128.09 g/mol
Boiling Pt:80…81 °C
Density:1.17 g/cm³ (25 °C)
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:208-075-3
UN:1987
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together