
4-(Trifluoromethyl)-2-pyridinecarbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:936841-69-9
:
Formula:C₇H₃F₃N₂
MDL Number:MFCD09038236
:172.11 g/mol
Boiling Pt:58…60 °C/1.5mmHg
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together