Order Entry
Czech Republic
ContactUsLinkComponent
2-(Trifluoromethyl)phenothiazine 97%
2-(Trifluoromethyl)phenothiazine 97%
Catalog # APOSPC7694-25G
Supplier:  Apollo Scientific
CAS Number:  
2-(Trifluoromethyl)phenothiazine 97%
Catalog # APOSPC7694-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :267.27 g/mol
Melting Pt:188…190 °C
Storage Temperature:Ambient
CAS Number:92-30-8
Formula:C₁₃H₈F₃NS
MDL Number:MFCD00005018