Order Entry
Czech Republic
ContactUsLinkComponent
3'-(Trifluoromethyl)formanilide 97%
Irritant
3'-(Trifluoromethyl)formanilide 97%
Catalog # APOSPC7670-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3'-(Trifluoromethyl)formanilide 97%
Catalog # APOSPC7670-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.14
 :GHS07
Storage Temperature:Ambient
CAS Number:657-78-3
Formula:C₈H₆F₃NO
MDL Number:MFCD00021013