
1-[4-(Trifluoromethoxy)phenyl]tetrahydro-2H-imidazol-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:162748-20-1
:
Formula:C₁₀H₉F₃N₂O₂
MDL Number:MFCD08689725
:246.19 g/mol
Melting Pt:145…146 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together