
5-Fluoro-2-methylindole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:399-72-4
:
Formula:C9H8FN
MDL Number:MFCD02093649
:149.17 g/mol
Boiling Pt:112…120 °C/0.1mmHg
Melting Pt:97…101 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together