

3-(Trifluoromethyl)benzylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2740-83-2
:
Formula:C₈H₈F₃N
MDL Number:MFCD00008117
Boiling Pt:93…97 °C/28mmHg
Density:1,222 g/cm³ (25 °C)
Flash Pt:71 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together