1-(tert-Butyl)-4-ethyl-5-(trifluoromethyl)-1H-pyrazole-1,4-dicarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1006492-02-9
:
Formula:C₁₂H₁₅F₃N₂O₄
MDL Number:MFCD08689692
:308.26 g/mol
Melting Pt:107…108 °C
Storage Temperature:Ambient
Frequently Bought Together