
2-(Bromomethyl)-4-(trifluoromethoxy)phenylboronic acid pinacol ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957066-13-6
:
Formula:C₁₄H₁₇BBrF₃O₃
MDL Number:MFCD09475917
:380.993 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together