
4-(Trifluoromethyl)benzophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:728-86-9
:
Formula:C₁₄H₉F₃O
MDL Number:MFCD00000400
Melting Pt:114…119 °C
:250.22 g/mol
EINECS:211-974-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together