
3-(Trifluoromethyl)benzophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:728-81-4
:
Formula:C₁₄H₉F₃O
MDL Number:MFCD00000389
:250.22 g/mol
Boiling Pt:121…123 °C/1.5mmHg
Melting Pt:51…52 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:211-973-8
:GHS07
Frequently Bought Together