Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Catalog # APOSPC7557-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Catalog # APOSPC7557-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :303.28 g/mol
Storage Temperature:Ambient
CAS Number:1000339-78-5
Formula:C₈H₈F₃NO₄S₂
MDL Number:MFCD09750912