
α,α,α-Trifluoro-o-toluonitrile 98%
Catalog # APOSPC7550-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:171.12 g/mol
Boiling Pt:204…206 °C
Melting Pt:17…19 °C
Density:1.294 g/cm³ (25 °C)
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:207-184-3
UN:3439
ADR:6.1,III
:GHS07
CAS Number:447-60-9
Formula:C₈H₄F₃N
MDL Number:MFCD00001791