
5-Bromomethyl-2-methyl-4-trifluoromethyl-1,3-thiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000339-73-0
:
Formula:C₆H₅BrF₃NS
MDL Number:MFCD09264554
:260.08 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together