4-(Trifluoromethylthio)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4021-50-5
:
Formula:C₈H₅F₃OS
MDL Number:MFCD00129191
:206.19 g/mol
Boiling Pt:219 °C
Melting Pt:29…31 °C
Density:1.31 g/cm³ (25 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
Frequently Bought Together