
4-(Trifluoromethoxy)phenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64285-95-6
:
Formula:C₈H₄F₃NOS
MDL Number:MFCD00042412
:219.19 g/mol
Boiling Pt:215…218 °C
Density:1.35 g/cm³ (25 °C)
Flash Pt:107 °C
Storage Temperature:Refrigerator
UN:1760
ADR:8,II
:GHS05
Frequently Bought Together